Super Erecta Shelf® Solid Shelving, Galvanized, Metro™
Supplier: Metro
2124LG 2448FG 1442FG 1460FG 1842FG 1860FG 2124FG 2130FG 2136FG 2142FG 2148FG 2430FG 2460FG 1436LG 1442LG 1448LG 1460LG 1824LG 1836LG 1842LG 1860LG 2130LG 2142LG 2148LG 2430LG 2442LG 2460LG 1836FG 2424FG 1448FG 1848LG 2448LG 2436FG 1436FG 1830FG 2160FG 1424LG 1430LG 2136LG 2424LG 2436LG 1830LG 2160LG 1824FG 1424FG 1430FG
97005-604EA 240.16 USD
97005-604 97005-820 97005-376 97005-380 97005-384 97005-386 97005-388 97005-390 97005-392 97005-394 97005-396 97005-400 97005-404 97005-584 97005-586 97005-588 97005-590 97005-592 97005-596 97005-598 97005-602 97005-606 97005-610 97005-612 97005-618 97005-622 97005-626 97005-812 97005-816 97005-378 97005-600 97005-624 97005-818 97005-374 97005-382 97005-398 97005-580 97005-582 97005-608 97005-616 97005-620 97005-594 97005-614 97005-810 97005-814 89369-712 89369-714
Super Erecta Shelf® Solid Shelving, Galvanized, Metro™
Furniture Shelves
Solid shelving features 18-gauge galvanized steel construction with a clear, baked-on acrylic coating, uncoated aluminum cast corners, and flat, embossed, or louvered/embossed surfaces.
  • Sturdy aluminum castings firmly lock the corners to the posts (sold separately)
  • Each post has rolled, circular grooves along its entire length at 2.5 cm (1") intervals
  • Unique tapered split sleeve (plastic or aluminum) snaps together around each post
  • Tapered openings in the shelf corners slide over the tapered split sleeves, providing a positive lock
  • Shelf is assembled in minutes without the use of nuts, bolts, or special tools
Two-fold thickness forms a 3.2 mm (¹/₈") raised ship's edge on all four sides for all-around rigidity and high strength
Stationary posts over 189.6cm high and mobile posts over 138.6 cm high are not for use with 35.6 cm wide shelves. The height of the chrome-plated and stainless steel stationary posts includes a leveling bolt and cap. The 245.4 cm high stationary posts are not for use with shelves less than 61 cm wide.
Ship's edges help contain spillage, mimimize contamination, and permit easier cleanup. Louvered/embossed shelf designs allow air circulation under packages and containers. Shelves can be loaded and unloaded from all sides for maximum use of storage space. Adjustable feet (bolt levelers) compensate for surface irregularities.
Ordering information: Stability decreases as the ratio of height to width increases; units should be kept as wide and low as possible. Each shelf and post is sold separately. For casters used with mobile units, see 19766-236 series. For special posts used to create shelving units with counter tops, see 19766-814 series. For other accessories, contact your Avantor representative.