Boekel Mini Vortex Mixer, Tap Dancer
Supplier: Boekel
270100
97004-374EA 390.94 USD
97004-374
Boekel Mini Vortex Mixer, Tap Dancer
Shakers and Mixers Vortex Mixers
Mixer features compact, with low profile and small footprint
Pressure sensitive cup and continuous or touch operation are also features of the vortex mixer. Rubber suction pads prevent walking, absorb vibration. Low voltage supply for safe cold room use. Two year warranty included.