Orbital and Reciprocating Shaking Water Bath, 230 V
Supplier: Boekel
290400-2
97004-348EA 7533.57 USD
97004-348
Orbital and Reciprocating Shaking Water Bath, 230 V
Baths Shaking Water Baths
The Boekel Scientific Orbital and Reciprocating Shaking Water Bath is designed for a wide range of temperature-controlled shaking protocols, such as northern and southern hybridization, destaining of gels, incubation of cells or bacterial cultures as well as wash processes.
  • 230 V model
The shaking water bath is equipped with a PID controller for easy temperature selection, rapid heat up and excellent stability. Choose from a wide variety of trays to maximize your versatility, such as a Universal Tray, test tube trays and a Perforated Tray. The perforated tray allows you to use the bath as a general purpose bath as well.
It features a high quality, robust design with a unique shaking mechanism that allows you to easily switch from orbital mode to reciprocating mode by simply reversing the tray. No tools needed. The convenient drain port with a quick disconnect allows for easy cleaning of the bath. The temperature range, which goes up to 99 °C also allows you to use the bath as a boiling bath.
Certifications: UL and CSA listed, CE Certified.
Ordering information: Unit comes equipped with a clear polycarbonate lid, power cord and instruction manual. All units include a two year warranty on parts and labor. Select a universal flask tray or a test tube tray to use with this bath. The universal flask tray and the test tube are sold seperately but required for use with this bath.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (97004-346) is also available, please see separate product page.