Orbital and Reciprocating Shaking Water Bath, Boekel Scientific
Supplier: Boekel
290400
97004-346EA 5590.35 USD
97004-346
Orbital and Reciprocating Shaking Water Bath, Boekel Scientific
Baths Shaking Water Baths
The Boekel Scientific Orbital and Reciprocating Shaking Water Bath is designed for a wide range of temperature-controlled shaking protocols, such as Northern and Southern Hybridization, Destaining of gels, Incubation of cells or bacterial cultures as well as Wash Processes
The Shaking Water Bath is equipped with a PID Controller for easy temperature selection, rapid heat up and excellent stability. Choose from a wide variety of trays to maximize your versatility, such as a Universal Tray, Test Tube Trays and a Perforated Tray. The Perforated Tray allows you to use the bath as a General Purpose Bath as well.
It features a high quality, robust design with a unique shaking mechanism that allows you to easily switch from orbital mode to reciprocating mode by simply reversing the tray. No tools needed. The convenient drain port with a quick disconnect allows for easy cleaning of the bath. The temperature range, which goes up to 99°C also allows you to use the bath as a Boiling Bath.
Certifications: UL and CSA listed, CE Certified.
Ordering information: Unit comes equipped with a clear polycarbonate lid, power cord and Instruction Manual. All units include a two year warranty on parts and labor. Select a Universal Flask Tray or a Test Tube Tray to use with this Bath. The Universal Flask Tray and the Test Tube are sold seperately but required for use with this Bath.