Pressure Transducer for Krosflo Pressure Monitor, Spectrum® Laboratories
Supplier: Spectrum Laboratories
ACPM-499-03N
97004-062PK 427.75 USD
97004-062
Pressure Transducer for Krosflo Pressure Monitor, Spectrum® Laboratories
Manometers
Polycarbonate and polysulfone single-use pressure transducers are designed to accurately measure feed, permeate, and retentate pressure during Tangential Flow Filtration process.