Autoclavable Carts and Accessories, Metro™
Supplier: Metro
LC2436AC LC2S1824 LC3S1824 LC3S2436 LC1824AC LC2S2436
97002-850EA 1014.14 USD
97002-850 97002-840 97002-842 97002-846 97002-848 97002-844
Autoclavable Carts and Accessories, Metro™
Carts
Lab carts include two or three stainless steel autoclave shelves with aluminum split sleeves and four stainless steel swedged caster posts
Ordering information: Autoclavable casters must be ordered separately. Additional autoclave shelves and aluminum split sleeves are also ordered separately.