LE510 Redox Combination Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
51340339
97002-312EA 218.39 USD
97002-312
LE510 Redox Combination Electrode, METTLER TOLEDO®
Electrodes Redox Electrodes
Gel-filled electrode is for use with METTLER TOLEDO® pH meters in water analysis.
  • Gell-filled
  • Plastic body
  • BNC connector
Robust plastic body makes it ideal for field measurements. Features BNC connector.