LE703 Four-Pole Graphite Conductivity Probe, METTLER TOLEDO®
Supplier: Mettler Toledo
51340335
97002-308EA 296.46 USD
97002-308
LE703 Four-Pole Graphite Conductivity Probe, METTLER TOLEDO®
Electrodes Conductivity Sensors
General-purpose probe is designed for use with METTLER TOLEDO® conductivity meters in a laboratory setting.
  • For conductivity testing
  • Four graphite poles
  • Plastic shaft
Probe features four graphite poles embedded into plastic shaft to eliminate polarization.