Heidolph™ Heat-On™ Heat-On Blocks
Supplier: Heidolph NA, LLC
015882020 015882030 015882050 015882060 015882070 015882100 015882120 015882130 015882180 015882190 015882230 015882270 015882280 015882290 015882300 015882310 015852000 015852010 015852020 015882010 015882040 015882220 015882250 015882260 015852030 015882000 015882080 015882160 015882170 015882210 015882240 015882090 015882110 015882200 015882055 015882056 015882580 015882105
97002-034EA 156.2 USD
97002-034 97002-036 97002-040 97002-042 97002-044 97002-050 97002-054 97002-056 97002-066 97002-068 97002-076 97002-084 97002-086 97002-088 97002-090 97002-092 97002-094 97002-096 97002-098 97002-032 97002-038 97002-074 97002-080 97002-082 97002-100 97002-030 97002-046 97002-062 97002-064 97002-072 97002-078 97002-048 97002-052 97002-070 10145-576 10145-578 10812-582 10812-584
Heidolph™ Heat-On™ Heat-On Blocks
Block Heaters Blocks
A safer, cleaner alternative to oil baths and heating mantles, blocks are for heating and stirring one to two round bottom flasks at a time, up to 200 °C (392 °F) or 260 °C (500 °F) 20 minutes or less.
Lightweight, solid aluminum blocks heat evenly and feature a deep well design for rapid heat transfer, so flasks will not stick or crack. Polymer or anodized coating provides strong chemical resistance. Blocks have two probe holes for temperature probe (not included), and fit on all common hotplates up to 145 mm in diameter.