InLab® Redox Micro Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
51343203
97001-694EA 530 USD
97001-694
InLab® Redox Micro Electrode, METTLER TOLEDO®
Electrodes Redox Electrodes
Electrode measures oxidation/reduction power of a solution
  • Platinum ring indicator
  • Safelock™ closing ring
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head connector
Features a platinum ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters. Diameter: 6mm.