InLab® Redox Ag Combination Electrode, METTLER TOLEDO®
Supplier: Mettler Toledo
51343205
97001-690EA 505 USD
97001-690
InLab® Redox Ag Combination Electrode, METTLER TOLEDO®
Electrodes Redox Electrodes
Electrode measures oxidation/reduction power of a solution
  • Silver ring indicator
  • Safelock™ closing ring
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head connector
Features a silver ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.