InLab® Cool Combination pH Electrode, Sleeve Junction, METTLER TOLEDO®
Supplier: Mettler Toledo
51343174
97001-646EA 650 USD
97001-646
InLab® Cool Combination pH Electrode, Sleeve Junction, METTLER TOLEDO®
Electrodes pH Electrodes
Electrode uses FRISOCOLYT-B™ electrolyte for low temperature samples
  • FRISOCOLYT-B™ electrolyte
  • Firm ground joint sleeve
  • ARGENTHAL™ lead-off and silver ion trap
  • S7 screw head
Features firm ground joint sleeve junction and ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connects with different cables, so electrode may be used with a wide range of meters. Supplied with 25mL electrolyte vial.