Reversing Digital Overhead Stirrer, Caframo
Supplier: Caframo
BDC2010 BDC2010 BDC2010U1 BDC2010U1 BDC2010AU
97001-136CS 10472.6 USD
97001-136 10860-290 CA97001-136 10860-016 CA10860-290
Reversing Digital Overhead Stirrer, Caframo
Stirrers Overhead Stirrers
The Reversing Digital stirrer maximizes productivity, fits into any area of the laboratory including a fume hood or enclosed workstation, and provides accurate and repeatable results every time.
This stirrer also offers a choice of rotation direction - clockwise, counter clockwise or both (agitation mode) and features a digital display, a countdown timer and a cycle timer.
Suitable for a wide range of mixing applications, this Caframo benchtop overhead stirrers are used in chemical reactions and synthesis, raw material QC, food sample prep, polymer and personal care formulations.
With the push of a button, the unique Stirlight™ lights up so that solutions can be seen clearly. The Reversing Digital Stirrer is powered by a brushless DC (BDC) motor. The belt driven transmission is smooth, quiet and maintenance free.
Ordering information: Clamp, stand, and impellers are available. Chuck (adjustable up to 10 mm or 3/8 in), chuck key, mounting support rod, external power supply, grounded power cord and manual in four languages (English, French, Spanish and German) are included. Three year warranty.