Clarity Urocheck 120 Urine Analyzer, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-FREEUAM DTG-UAM DTG-UAPROMO
89135-184EA 2298.25 USD
89135-184 95064-116 95064-118
Clarity Urocheck 120 Urine Analyzer, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Analysers
This urine strip reader saves time by testing up to 120 samples per hour, and provides results on a large, easy-to-read LCD display
Abnormal results are flagged. Features QC Lockout for administrator and operator protection, as well as automatic calibration and self-check for easy maintenance. The reader is compatible with all LIS, HIS, EMR, and HER systems and includes a 2000-test internal memory, USB and RS232 ports, and an optional bar code scanner
Certifications: Certifications: Analyzer and strips are CLIA waived.
Ordering information: Analyzer is available in kits that includeMore Product Information