Clarity FOB Controls, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-FOBCTLS
95064-014CS
4038.53
USD
95064-014
Clarity FOB Controls, Clarity Diagnostics
Clinical Diagnostic Supplies
Clinical Reagents and Kits
Can be used to validate Clarity IFOBT and other brand rapid IFOB tests. Contains 1 vial of positive and 1 vial of negative control.
- 1 ml/vial
- Room temperature storage
- 16-18 months dating at the time of purchase
Can be used as training tool.