Clarity Strep A Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-STP50 DTG-STP25 DTG-FLIP
95064-010CS 1931.47 USD
95064-010 95064-008 95064-012
Clarity Strep A Test, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
This kit is a rapid assay for qualitative detection of the Strep A antigen from throat swabs
Tests have 95% accuracy in rare and 1+ colonies versus culture tests and 99% specificity. Easy-to-read results are available in five minutes or less. Tests can be stored at room temperature for up to 24 months. Additional swabs and tubes are sold separately.
The kit contains all the materials needed to perform the test, including extraction reagents A and B, positive and negative controls, extraction tubes, throat swabs, and a reaction tube rack. Kits are available in strip or cassette format.
Certifications: CLIA waived