Clarity Mono Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
Clarity provides an easy-to-use diagnostic product that can quickly test for Mononucleosis. This CLIA Waived test is ideal for point-of-care testing that allows you to provide accurate results in no more than 8 minutes. Package contains reagent, controls, tubes, and cassettes for 15 tests.
- Sensitivity > 99%
- Specificity of 98.6%
- Fast and easy test results in 8 minutes or less
- No age restriction (can be used on children)
- 18 month dating
Adding this test to your practice has the potential to create a new revenue stream for your practice while making your facility more efficient at caring for patient’s needs.
Certifications: CLIA waived for whole blood only, can be used with plasma/serum.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...