Clarity Urine/hCG Liquid Control, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-UHCTLS
95064-002CS
1343.04
USD
95064-002
95064-000
Clarity Urine/hCG Liquid Control, Clarity Diagnostics
Controls and Calibration Verification Materials
Clarity Urine Controls for use with Siemens/Roche Urine Strips and Siemens/Roche Urine analyzers only.
- Dropper style controls
- Color coded cap: Blue cap for negative and red cap for positive
- No refrigeration required in transit
- Equivalent to BioRad and Quantimetrix urine controls
For use with Siemens and Roche Micro albumin creatine test strips. For use with all Rapid hCG/Pregnancy tests including Clarity hCG Tests.
Comes in easy-dispensing dropper bottles, and remains stable in a liquid state for two years, even after being opened.
Certifications: CLIA waived.