Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Supplier: Clarity Diagnostics
DTG-10SG DTG-5OB DTG-KET DTG-4OB DTG-2GP
95064-046CS 1023.12 USD
95064-046 95064-050 95064-056 95062-919 95064-055
Clarity Urocheck Urinalysis Strips, Clarity Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Test strips for urinalysis testing of various parameters.
  • Consistent, highly accurate and sensitive
  • Competitively priced
  • Can be read visually or by using the Clarity 120C Urine Analyzer
  • 16 to 18 month dating
  • 90 day open vial stability
  • Room temperature storage
  • Read results between 60 to 20 seconds
  • Color chart made in Germany
  • CAP/API values listed
Strips detect the following 10 parameters: Leukocytes/Nitrite/Urobilinogen/Protein/Ph/Blood/Specific Gravity/Ketone/Bilirubin/Glucose in urine samples.
Ideal for POL, Physician offices, Urgent Care Locations, OB-GYN offices, and Hospitals.
Tests strips have an 18-month shelf life.
Certifications: CLIA waived. FDA approved.
Packaging: 100 strips per bottle.