Tryptic Soy Broth Dehydrated Culture Media, BD Diagnostics
Supplier: BD
257488
95062-836PK 207.23 USD
95062-836
Tryptic Soy Broth Dehydrated Culture Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tryptic Soy Broth are nutritious media that will support the growth of a wide variety of microorganisms, including common aerobic and facultatively anaerobic bacteria and fungi.
It is used in qualitative procedures for the detection, isolation, and cultivation of fastidious and nonfastidious microorganisms, in sterility testing, and in the microbiological examination of nonsterile products.