Petri Dish Tissue Chambers, BTX™
Supplier: Harvard Apparatus
45-0506
95062-362EA 1688.94 USD
95062-362
Petri Dish Tissue Chambers, BTX™
Culture Dishes Cell Culture Dishes
This tissue chamber/electrode is specifically designed to handle ex-vivo tissue samples that are too large or have a unique shape that make it difficult to transfect using other standard electrodes
Transfection of ex-vivo tissue samples is an efficient method to deliver genes and drugs to a wide range of tissue types including cornea, muscle, and skin. With the use of this specially designed tissue sample chamber, transfection is made simple and easy. The chambers are available in varied widths; 5mm and 15mm to accommodate numerous tissue sample sizes. The reusable chamber consists of a laboratory grade Pyrex glass petri dish and two platinum electrodes embedded in an inert silicone, creating the rectangular chamber that provides a homogenous field of energy for high transfection efficiencies.
Ordering information: Tissue chamber 95062-360 includes a 5x5mm glass petri dish and micro grabber connection cables. Tissue chamber 95062-364 includes a 15x15mm glass petri dish and micro grabber connection cables. Tissue chamber electrode 95062-362 includes a 15x15mm tissue chamber electrode (connection cables must be ordered separately).