Sabouraud Dextrose Agar with Lecithin and Polysorbate 80, bioMerieux
Supplier: bioMerieux
P2820
95061-514CS 594.18 USD
95061-514
Sabouraud Dextrose Agar with Lecithin and Polysorbate 80, bioMerieux
Media for Microbiology Prepared Media for Microbiology
Sabouraud Dextrose Agar w/Lecithin and Polysorbate 80 is used for the detection and enumeration of viable fungal contaminants present on surfaces and equipment of sanitary importance.
  • For isolation and cultivation of fungi from surfaces sanitized with quaternary ammonium compounds
  • 10 x 500 mL
Upon receipt, store plates at 2 – 8 °C away from direct light in an inverted position. Store bottles according to the package label in an upright position. Store plates in original outer wrapping until ready to use. Media can be inoculated up to the expiration date and incubated for the appropriate incubation period.