Fluid B Rinsing Solution, Biomerieux
Supplier: bioMerieux
P2750-RS
95061-502CS 272.69 USD
95061-502
Fluid B Rinsing Solution, Biomerieux
Microbiology Test Systems Endotoxin Tests
Sterile diluent for rinsing membranes during sterility testing by membrane filtration on articles containing lecithin or oil recommended by USP/EP/JP.
  • Effective
  • Large quantity of solution
Rinses membranes during sterility testing by membrane filtration on articles containing lecithin or oil recommended by USP/EP/JP.