API® OF Medium Microbial Identification Kit, Biomerieux
Supplier: bioMerieux
50110
95060-850PK 76.48 USD
95060-850
API® OF Medium Microbial Identification Kit, Biomerieux
Microbiology Test Systems Microorganism Tests
This medium is for use as a reagent with API® test strips for the determination of fermentation or oxidation metabolism.
  • Economical
  • Easy to use
  • Effective reagent
OF medium is recommended for the detection of oxidation or fermentation of carbohydrates by bacteria.