QUANTISWAB® Irradiated Swabs for Media
Supplier: bioMerieux
43801
95060-840CS 1583.94 USD
95060-840
QUANTISWAB® Irradiated Swabs for Media
Swabs Swabs for Sampling
QUANTISWAB® is an irradiated, triple-wrapped environmental swab utilizing nylon-flocked technology. This patented technology offers the QUANTISWAB® a unique design allowing up to 60% recovery of micro-organisms from surfaces compared to a recovery rate of 20% for traditional swabs.
  • Ideal for sampling of irregular surfaces
The QUANTISWAB® has also demonstrated a 90% release capacity of microorganisms captured by the swab.
Specifically designed for aseptic manufacturing facilities, QUANTISWAB® product is protected by triple sterile packaging, perfect for the biopharmaceutical and cosmetic industries.
Ordering information: Made to order items have a lead time of 42 days.