API® 20 Microbial Identification Kits, bioMerieux
Supplier: bioMerieux
20300
20100
20160
20600
20701
20050
95060-682PK
869.53
USD
95060-682
95060-674
95060-676
95060-686
95060-688
95060-670
API® 20 Microbial Identification Kits, bioMerieux
Microbiology Test Systems
Microorganism Tests
Standardized, miniature version of conventional methods conducting various microbial tests.
- Standardized
- Easy to use
- Identify gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other microorganisms
These microbial identification kits are used for identifying gram-negative, gram-positive, and anaerobic bacteria, yeasts, and other clinically significant microorganisms.
Additional reagents required see package insert for details
https://us.vwr.com/store/product/18630970/microbial-identification-reagents-biomerieux
API Web software required for Identification -purchase directly from bioMerieux.