Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
Supplier: THERMO CLINICAL CHEMISTRY
100600
95059-818PK 682.3 USD
95059-818
Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
Clinical Diagnostic Supplies Clinical Reagents and Kits
These assays are used to screen and quantify plasma fibrinogen based on the Clauss method
They are useful in screening for fibrinogen disorders by measuring the formation of the fibrin clot, and detect acquired and congenital hemorrhagic disorders. The tests have a reconstituted stability of 8 hours when stored at 2 to 8°C (36 to 46°F). Test components are also available separately.