Normal Goat Serum, KPL
Supplier: KPL
5560-0007
95059-444EA 334.53 USD
95059-444
Normal Goat Serum, KPL
Sera (Serum)
Normal Goat Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen.
  • Provides an alternative to bovine serum albumin (BSA)
  • Reduce non-specific binding of proteins
  • Maximum signal-to-noise ratio
Normal Goat Serum can be used as a blocking agent to treat plastic surfaces, membrane or tissue after they have been sensitized with primary antibody or antigen. It provides an alternative to bovine serum albumin (BSA) and non-fat dry milk. It is effective in reducing non-specific binding of proteins to reaction surfaces, thereby maximizing signal-to-noise ratio. This blocking agent is recommended for use in immunoassays where the primary antibody was produced in goat, as a source of non-specific serum protein or on tissue for immunohistochemical applications.