GeneRuler™ Biotinylated DNA Ladder, KPL INC
Supplier: KPL
5960-0029
95059-388EA 1179.53 USD
95059-388
GeneRuler™ Biotinylated DNA Ladder, KPL INC
Electrophoresis Reagents Molecular Weight Markers
The GeneRuler™ Biotinylated DNA Ladder is a convenient prelabeled molecular weight marker ideal for use on southern blots.
  • 21 distinct bands are visible from 100 bp-10 kb
  • Bright bands at 3000 bp, 1031 bp, and 500 bp serve as reference for easy orientation
The ladder contains a mixture of labeled and unlabeled DNA which migrate uniformly and will not interfere or cross hybridize with the target.