Tris Glycine Transfer Buffer, 10X, Seracare
Supplier: KPL
5460-0011
95059-240EA 150.16 USD
95059-240
Tris Glycine Transfer Buffer, 10X, Seracare
Buffers Electrophoresis Buffers Transfer Buffers
Tris-glycine transfer buffer is ideal for preparing a standard western blot transfer buffer (Towbin) and for use as a gel electrophoresis buffer for native Tris-glycine gel without SDS.
  • pH=8.3
Conductivity controlled.