Milk Diluent/Blocking Concentrate Kit, KPL Immunoassay Kits, Seracare
Supplier: KPL
5140-0011
95059-192EA 322.84 USD
95059-192
Milk Diluent/Blocking Concentrate Kit, KPL Immunoassay Kits, Seracare
Buffers Electrophoresis Buffers Blocking Buffers
Convenient antibody diluent/blocking solution concentrate containing 2% nonfat milk in a borate buffer.
Recommended for blocking membranes in western blots or as an alternative blocker in microwell ELISA.