Anti-IgG gamma Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Supplier: KPL
5220-0348
95058-750EA 513.49 USD
95058-750
Anti-IgG gamma Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antibodies
Secondary Goat Anti-IgG(gamma) Reacts with Human

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR