Anti-IgG Goat Polyclonal Antibody (DyLight® 488)

Supplier: KPL
5230-0391
95058-512EA 327.76 USD
95058-512
Anti-IgG Goat Polyclonal Antibody (DyLight® 488)
Antibodies
Secondary Goat Anti-IgG (H+L) Reacts with Mouse

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR