Hematoxylin solution Premium, VWR® stain for histology

Supplier: VWR International

95057-844
95057-844CS 1381.36 USD
95057-844
Hematoxylin solution Premium, VWR® stain for histology
Hematoxylin

Premium histology stains and reagents are designed to complement one another to provide optimum results, enabling greater control of slide presentation when used together.


  • Analytical chemistry standards and performance tests are applied to ensure consistency
  • Complementary use provides an excellent value for the cost


Each stain and reagent is chemically engineered to support the others, resulting in reliably consistent results from the first slide to the last


Hematoxylin provides extra sharp nuclear detail with notably fast staining times for increased productivity in the lab. This remarkably consistent stain offers a wide range of darkness, allowing pathologists the ability to obtain the desired personal shade, color, and intensity.


All stains and reagents undergo performance tests and pass analytical chemistry standards.

Formula: C₁₆H₁₄O₆
Storage Temperature: Ambient
MDL Number: MFCD00078111
CAS Number: 517-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR