DNA Ladders, G-Biosciences
Supplier: G-Biosciences
786-462
95057-692EA 135.1 USD
95057-692
DNA Ladders, G-Biosciences
Electrophoresis Reagents Molecular Weight Markers
G-Biosciences' 1.0 kb DNA ladder is a ready-to-use preparation that consists of 15 DNA fragments ranging from 100 bp to 10 kb
G-Biosciences' 500 bp DNA ladder is a ready-to-use preparation that consists of 10 DNA fragments ranging from 500 bp to 5.0 kb. The recommended loading is 5 μL DNA marker per lane. The 500 bp ladder can be used for approximating the mass of DNA in comparably intense samples of similar size.
G-Biosciences' 100 bp DNA marker is a ready-to-use preparation that consists of 14 fragments, ranging form 100bp to 3,000 bp. The recommended loading is 5 μL DNA marker per lane. The 100 bp ladder cannot be used for precise quantification of DNA mass, but can be used for approximating the mass of DNA in comparably intense samples of similar size.
G-Biosciences' 50 bp DNA marker is a ready-to-use preparation that consists of 13 fragments, ranging form 50 bp to 1,000 bp. The recommended loading is 5 μL DNA marker per lane. The 50 bp ladder cannot be used for precise quantification of DNA mass, but can be used for approximating the mass of DNA in comparably intense samples of similar size.
The recommended loading is 5 μL DNA marker per lane. The 1.0 kb ladder can be used for approximating the mass of DNA in comparably intense samples of similar size.
Ordering information: Accessories available.