Immunohistochemistry Staining Trays, IBI Scientific
Supplier: IBI Scientific
AT-12 ATC-12 AT-24 ATC-24
95039-740EA 380.74 USD
95039-740 95039-742 95057-478 95057-480
Immunohistochemistry Staining Trays, IBI Scientific
Staining Boxes
Designed specifically for the incubation and storage of immunohistochemistry slides, these specially machined trays securely hold up to 12 or 24 slides in place, and the built-in reservoir allows water to be added during incubation
Lids are available in black or clear cast acrylic. Black lid shields light-sensitive slides, eliminating the need to wrap staining trays in aluminum foil.
Trays stack easily and securely on top of each other for a neat and tidy benchtop.
Weight: 12-Slide Assay Staining Tray: 0.9 kg (2 lbs.), 24-Slide Assay Staining Tray: 1.8 kg (4 lbs.)