VACUETTE® Heparin Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
454247P 456087P 454237 454029 454008P 456088 455084 454030 456028 455051 454244
95057-213CS 377.88 USD
95057-213 95057-217 95057-219 95057-399 95057-403 95057-409 95057-215 95057-405 95057-407 95057-411 95057-413 95057-415 95057-401
VACUETTE® Heparin Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Tubes feature an inner wall coated with spray-dried sodium heparin or lithium heparin.
  • Tubes contain anticoagulant
  • Color-coded rings in caps
  • PET plastic construction is virtually unbreakable
The anticoagulant heparin activates antithrombins, which block the coagulation cascade and produce a whole blood/plasma sample rather than clotted blood plus serum
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with green non-ridged pull caps that feature an exterior color-coded ring to indicate either the additive used or draw volume. A black ring indicates standard tubes, a yellow ring indicates the inclusion of a separation gel, and a white ring indicates a tube designed to accommodate pediatric draw.
Tubes are also available with lithium heparin coating and a gel separator. The specific gravity of the gel lies between the blood cells and plasma so that, during centrifugation, the gel barrier moves to form a stable barrier separating plasma from cells. Plasma may then be aspirated directly from the collection tube, eliminating the need for transfer to another container.