VACUETTE® Trace Element Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
456275
95057-259CS 1481.3 USD
95057-259
VACUETTE® Trace Element Tubes, Greiner Bio-One
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
VACUETTE® trace element tubes are designed to test trace elements such as aluminum, lead, cadmium, chrome, iron, fluoride, cobalt, copper, lithium, manganese, molybdenum, mercury, selenium, or thalium.
  • With sodium heparin as additive
  • Color-coded rings in caps indicate draw volume
  • PET plastic construction is virtually unbreakable
VACUETTE® tubes are manufactured from virtually unbreakable PET plastic. These clear tubes are supplied with royal blue non-ridged pull caps that feature a black exterior color-coded ring.