Protein A HP SpinTrap Spin Columns, Cytiva

Supplier: Cytiva
28-9031-32
95056-280PK 331.68 USD
95056-280
Protein A HP SpinTrap Spin Columns, Cytiva
Filters Centrifugal Filters
Protein A HP SpinTrap™ columns are designed for the enrichment of proteins of interest, immunoprecipitation and the purification of monoclonal and polyclonal antibodies from cell lysates and biological fluids.

  • For simple, small-scale antibody purification or enrichment of specific proteins in prepacked, single-use spin columns that are ready to use
  • Classic and cross-link protocols provide flexibility
  • Elution conditions formatted for both electrophoresis and LC-MS analysis workflows.
  • Fast binding kinetics and high capacity provide high yield

Order Now

SPECIFICATIONS

Sample Volume 600 µl
pH Range 3 - 9
Storage Conditions 2 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR