GSTrap 4B Columns
Supplier: Cytiva
28-4017-48 28-4017-45 28-4017-47 29-0486-09
95055-968PK 2060.81 USD
95055-968 95056-278 95056-050 89501-374
GSTrap 4B Columns
Chromatography Columns
GSTrap 4B columns are prepacked Glutathione Sepharose 4B columns for convenient, high capacity one-step purification of glutathione S-transferase (GST) tagged proteins.
  • High binding capacity
  • Mild elution conditions preserving protein antigenicity and function
  • Easy one-step purifications of Glutathione S-Transferase (GST) tagged proteins resulting in high purity
  • GSTrap 4B are designed to be used with a syringe, pump, or chromatography system, such as ÄKTA design
GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap 4B. GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione. The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids.
GST-tagged proteins can be detected using colorimetric or immunological methods. The resin, Glutathione Sepharose 4B, is also available as lab packages and is a good choice for scale-up. The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System.