Ab Buffer Kit, Cytiva
Supplier: Cytiva
28-9030-59
95056-236EA
236.4
USD
95056-236
Ab Buffer Kit, Cytiva
Buffers
Antibody Purification Buffers
Ab Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of IgG according to recommended protocols.
The kit also includes neutralizing buffer. Ab Buffer Kit eliminates time consuming buffer preparation, which allows fast, reproducible, and convenient purification work.
Delivery information: Includes 50 ml binding buffer (10X stock solution), 15 ml elution buffer (10X stock solution), and 25 ml neutralizing buffer.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)