Sepharose™ CL-2B Gel Filtration Base Matrix
Supplier: Cytiva
17-0140-01
95056-222EA 1915.63 USD
95056-222
Sepharose™ CL-2B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose CL-2B is a well-proven cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 2% cross-linked agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability