Sepharose™ 4B Gel Filtration Base Matrix
Supplier: Cytiva
17-0120-01
95056-138EA 1198.73 USD
95056-138
Sepharose™ 4B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose™ 4B is an agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
  • 4% agarose gel filtration media
  • Proven base matrix for coupling affinity ligands