Sepharose™ CL-4B Gel Filtration Base Matrix
Supplier: Cytiva
17-0150-01
95056-028EA 1285.02 USD
95056-028
Sepharose™ CL-4B Gel Filtration Base Matrix
Protein Purification Protein Purification Resins
Sepharose™ CL-4B is a cross-linked agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix.
  • 4% cross-linked agarose gel filtration media
  • Proven base matrix for coupling affinity ligands
  • Higher degree of crosslinking improve matrix stability
The matrix is not pre-activated and the user performs all steps in coupling.