PD SpinTrap G-25 Columns, Cytiva
Supplier: Cytiva
28-9180-04
95055-982PK 485.77 USD
95055-982
PD SpinTrap G-25 Columns, Cytiva
Chromatography Columns
PD SpinTrap G-25 is a single-use column for cleanup of large biomolecules.
  • Convenient and fast small-scale cleanup of proteins and other large biomolecules (>5000 Mr) with the possibility to run multiple samples in parallel
  • Range of applications such as desalting, buffer exchange, and removal of low-molecular weight compounds
  • High desalting capacity
  • Reproducible, high throughput, parallel cleanup of protein samples
  • Sample preparation in the range of 100 to 180 μl without sample dilution
PD SpinTrap G-25 is a single-use microspin column that is designed for desalting and buffer exchange of biological sample using a standard microcentrifuge. The column is prepacked with ~600 µL Sephadex G-25 resin, and the column material is biocompatible polypropylene.
Delivery information: Instructions supplied with the columns.