Dextrin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Supplier: Cytiva
28-9355-98 28-9355-97
95055-918EA 4247.73 USD
95055-918 95055-916
Dextrin Sepharose™ High Performance Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Dextrin Sepharose™ High Performance is an affinity media for purification of recombinant proteins tagged with Maltose Binding Protein (MBP).
  • The small, evenly sized beads ensure that MBP-tagged proteins elute in narrow peaks, thus minimizing the need for further concentration steps
  • Physiological conditions and mild elution preserve target protein activity
  • Dextrin Sepharose High Performance tolerates all commonly used aqueous buffers and is easily regenerated using 0.5 M NaOH allowing the same column to be used for repeated purifications
Dextrin Sepharose High Perfromance is an affinity media for purification of proteins tagged with Maltose Binding Protein (MBP). Dextrin Sepharose High Performance and provide convenient affinity purification of recombinant proteins tagged with maltose binding protein (MBP). This robust and stable Sepharose High Performance based medium enables elution of target protein in narrow peaks, which minimizes the need for further concentration steps.