HiPrep Sephacryl S-200 HR, Cytiva
Supplier: Cytiva
17-1195-01 17-1166-01
95055-808EA 1409.56 USD
95055-808 95056-002
HiPrep Sephacryl S-200 HR, Cytiva
Chromatography Columns
HiPrep™ 16/60 Sephacryl® S-200 HR and HiPrep™ 26/60 Sephacryl® S-200 HR are pre-packed gel filtration columns for preparative separation of biomolecules of up to 5 and 13 ml sample volume respectively. Sephacryl® S-200 is used for separating mid-size proteins including antibodies and serum proteins.
  • Maximum recovery and minimum non specific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Columns provides reproducible separations over a wide molecular weight range