HiPrep Sephacryl S-100 HR, Cytiva
Supplier: Cytiva
17-1194-01 17-1165-01
95055-806EA 1390.85 USD
95055-806 95056-000
HiPrep Sephacryl S-100 HR, Cytiva
Chromatography Columns
HiPrep Sephacryl S-100 HR are prepacked gel filtration columns for preparative separation of biomolecules from up to 13 ml sample volume.
  • Maximum recovery and minimum nonspecific adsorption with long-term chemical and physical stability resulting from the hydrophilic, rigid allyl dextran/bisacrylamide matrix
  • Gel filtration columns for low to medium pressure ÄKTA systems.
  • Well-established in industrial production
  • The HiPrep Sephacryl series offers reproducible separations over a wide molecular weight range
  • Convenient, prepacked HiPrep Sephacryl columns speed up protein purification
Sephacryl S-100 is used for separating peptides and small proteins.