HyClone™ AdvanceSTEM Dulbecco's Phosphate Buffered Saline (DPBS), ES Cell Qualified, Cytiva
Supplier: Cytiva
A balanced salt solution tested for use with embryonic stem cells.
- Without calcium and magnesium
- 0.1 µm filter sterilised
- Storage temperature of 15 to 30 °C
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Caution: For further manufacturing or research use. Not for diagnostic or therapeutic use.