3,3',5,5'-Tetramethylbenzidine (TMB) substrate

Supplier: Enzo Life Sciences

Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine

ADI-80-1805
95052-796EA 105.73 USD
95052-796
3,3',5,5'-Tetramethylbenzidine (TMB) substrate
3,3',5,5'-Tetramethylbenzidine (TMB)

HRP substrate.


  • Used in ELISA


This TMB Substrate is intended for use as a colorimetric HRP-substrate for immunoassay development. Each bottle is sufficient for use with 5x96 wells microtiter plates.

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007748
CAS Number: 54827-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR